Chemical process indicators

Lieferant: CROSSTEX
CPI-E01 CPI-E02 SP-E16
NAMSCPI-E01EA 212 EUR
NAMSCPI-E01 NAMSCPI-E02 NAMSSP-E16
Chemical process indicators
Sterilisationsindikatoren
Chemical process indicators provide a fast and consistent visual check on items that were sterilised by EO.

  • Consistent and irreversible colour transition
  • Permanent pressure-sensitive adhesive
  • Indicator inks do not contain lead or other toxic heavy metals

Chemical indicator: Initial colour (purple) and signal colour (green)

Storage condition: 15 to 30 °C

Relative humidity: 20 – 70%

Standards: Compliance to ISO 11140-1:2014.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR