Mini spore discs
Lieferant: CROSSTEX
DN18-06
NAMSDN18-06EA 510 EUR
NAMSDN18-06
Mini spore discs
Sterilisationsindikatoren
Inoculated carrier mini spore discs for monitoring EO and dry heat processes.
  • An inoculated carrier, 3 mm disc, of Bacillus atrophaeus (cell Line 9372)
  • Primary packaging in bulk
The mini spore discs are designed to be placed directly into a device and utilised to monitor EO and dry heat sterilisation process efficacy.
The mini spore discs are labelled for industrial use only.
Standards: ISO 11138-1 Sterilisation of health care products – Biological indicators – Part 1: General requirements.
ISO 11138-2 Sterilisation of health care products – Biological indicators – Part 2: Biological indicators for ethylene oxide sterilisation processes.