Digital dry block incubator
Lieferant: CROSSTEX
INC-13E INC-13U
NAMSINC-13EEA 587 EUR
NAMSINC-13E NAMSINC-13U
Digital dry block incubator
Brutschränke Kleinbrutschränke
The digital dry block incubator is designed for convenient and reliable incubation of 1 ml spore ampules, SporView® SCBIs, SporView®10 SCBIs, and traditional SCBIs.
  • The electronically-heated aluminum block holds up to 10 units
  • Temperature and incubation time are displayed digitally
  • Reaches the set point temperature in 15 minutes or less
  • Transparent lid included to ensure consistent temperature and to protect the units