Spore discs, steel
Lieferant: CROSSTEX
TTS-06
NAMSTTS-06EA 829 EUR
NAMSTTS-06
Spore discs, steel
Sterilisationsindikatoren
Spore discs are used to monitor the plasma/vaporised hydrogen peroxide sterilisation processes. Place the spore discs either directly into the chamber or positioned in areas of a device which are the most difficult to sterilise or distributed throughout rooms and isolators.
  • An inoculated carrier, with a population level of 10⁶ spores per disc
  • 19-month shelf life when stored under labelled conditions
Standards: Meet USP and ISO 11138-1 standards.
Verpackung: Packed in easy-to-access Tyvek®/Tyvek® pouches.