Lieferant: NATIONAL INSTRUMENTS
783368-01
NATI783368-01EA 0 EUR
NATI783368-01
Datenlogger
GPIB-USB-HS+ IEEE 488 controller and portable analyser device for computers with USB slots. Users can use this device to integrate an instrument into their system using GPIB as well as to troubleshoot and solve GPIB hardware and software problems. The GPIB-USB-HS+ achieves maximum IEEE 488.2 performance. With no GPIB cable required for instrument connection, the user can use the Hi-Speed USB port to control up to 14 programmable GPIB instruments.
  • 61,875 Mbps data rate
  • Bus connector type is USB 2.0
The improved design allows for size compatibility with almost every instrument, adds analyser functionality, and reduces the overhead to generate faster turnaround times in your application. The device includes a license for the NI-488.2 driver software, providing maximum reliability for connecting to third-party instruments with GPIB.