Kryoboxen
Lieferant: NEOLAB
2-2701 2-2700
NEOL2-2701EA 5.07 EUR
NEOL2-2701 NEOL2-2700
Kryoboxen
Boxen Kryoboxen
These cryoboxes are made of cryo resistant and waterproof cardboard.
  • Lid has on the foreside a noticeable labelling pad
For storage of cryovials.