Labelling tapes, rainbow pack, NeoTape®
Lieferant: NEOLAB
2-6240
NEOL2-6240EA 190 EUR
NEOL2-6240
Labelling tapes, rainbow pack, NeoTape®
Klebebänder
These universal labelling adhesive tapes are assembled in Rainbow Pack with dispenser. The pack includes 10 coloured tape rolls (white, yellow, green, red, pink, blue, rosé, violet, lavender) of 13 mm width and 12,7 m length.
  • A multi-tape dispenser is included