Variocar Basic Trolley with 4 Drawers
Lieferant: NEOLAB
2-7023
NEOL2-7023EA 624 EUR
NEOL2-7023
Variocar Basic Trolley with 4 Drawers
Transportwagen
Mobile side table with worktop, drawers and storage compartment at the bottom.
  • Chrome-plated square tubular steel frame
  • Tops and drawers with high-quality, five-layer coating on both sides
  • 4 smooth-running castors with 75 mm Ø
  • Total load up to approx. 50 kg
  • Material: Wood/melamine
Drawers with smooth running telescopic ball bearing slides with locking mechanism.
The trolley is delivered disassembled, but can be easily assembled.
Frame material: Steel/chrome plated
Rollers material: Polycarbonate
Trolley: Transport/support/storage table trolley
Transport temperature: Room temperature