Shelf trolleys
Lieferant: NEOLAB
2-7041
NEOL2-7041EA 497 EUR
NEOL2-7041
Shelf trolleys
Transportwagen
The small shelf trays with square tubular steel frame are suitable for use in small devices or narrow rooms. The shelves comes with melamine resin coating on all sides which is resistant against disinfectants.
  • Rolls: four (50 mm Ø each)
  • Load capacity: ∼up to 20 kg
Construction: Steel frame, white varnished.