Transport trolley
Lieferant: NEOLAB
2-7083
NEOL2-7083EA 437 EUR
NEOL2-7083
Transport trolley
Transportwagen
Transport trolley made of plastic-coated steel tube, with two baskets.
  • Sturdy
  • Upper basket of 65 L and lower basket with 24 L capacity
  • 4 wheels, Ø 100 mm
Basket dimension: 600×400 mm