Jars, wide neck, with screw caps
Lieferant: NEOLAB
3-2129
NEOL3-2129EA 244 EUR
NEOL3-2129
Jars, wide neck, with screw caps
Dosen Dosen, Universal
High-quality jars, made of high-grade steel V4A, with polished surfaces.
  • Even water-proofed without sealing
  • Seamless interior without edges and corners
  • Easy to clean