Reaktionsgefäße
Lieferant: NEOLAB
7-4590
NEOL7-4590EA 89.3 EUR
NEOL7-4590
Reaktionsgefäße
Röhrchen Reaktionsgefäße
PP, amber coloured.
  • Ungraduated
  • Amber coloured to protect light sensitive substances
  • Excellent chemical resistance
  • Excellent temperature resistance
With thread.
Bestellinformation: Screw caps must be ordered seperately.