Thread Adapter, GL 32
Lieferant: NEOLAB
E-2933
NEOLE-2933EA 45.7 EUR
NEOLE-2933
Thread Adapter, GL 32
Fittings und Verbinder
These are required to attach threaded vials to ground joint apparatus such as rotary evaporators.
Bestellinformation: Size-dependent screw couplings are required for this purpose.