Bulb pipettes, one mark, Class B
Lieferant: NEUBERT VOLUME GLASSWAERE
0272-30-200
NEUB0272-30-200EA 60.5 EUR
NEUB0272-30-200
Bulb pipettes, one mark, Class B
Pipetten Vollpipetten
Glass.
  • Calibrated to deliver 'EX'
  • Full delivery
  • Blue graduations
Standards: DIN 12691