Graduated pipettes, Class AS, short form
Lieferant: NEUBERT VOLUME GLASSWAERE
0273-00-025
NEUB0273-00-025EA 31.4 EUR
NEUB0273-00-025
Graduated pipettes, Class AS, short form
Pipetten Vollpipetten
Short glass pipettes with wide opening (3 mm).
  • Calibrated to contain "EX"
  • Blue graduations