Volumetric flasks for crude oil analysis, glass, clear
Lieferant: NEUBERT VOLUME GLASSWAERE
0378-70-100190 0378-70-250 0378-70-087 0378-70-088 0378-70-089
NEUB0378-70-100190EA 144 EUR
NEUB0378-70-100190 NEUB0378-70-250 NEUB0378-70-087 NEUB0378-70-088 NEUB0378-70-089
Volumetric flasks for crude oil analysis, glass, clear
Kolben Messkolben
Graduated trapezoidal shaped flasks of borosilicate glass 3.3 with glass stoppers.
Used for the determination of the acid-separable fraction of water-miscible cooling lubricants.
Standards: DIN 51368