Centrifuge tubes
Lieferant: NEUBERT VOLUME GLASSWAERE
0474-31-2411
NEUB0474-31-2411EA 49.4 EUR
NEUB0474-31-2411
Centrifuge tubes
Röhrchen Zentrifugenröhrchen
Borosilicate glass 3.3.
  • Strong walled, without screw cap
  • With thread
Standards: According to DIN 58970 (Part II).