Cold traps, with olives, UT core
Lieferant: NEUBERT VOLUME GLASSWAERE
07257829402300 0725-80-29250 0725-80-45
NEUB07257829402300EA 78.5 EUR
NEUB07257829402300 NEUB0725-80-29250 NEUB0725-80-45
Cold traps, with olives, UT core
Vakuumfallen
The cooling traps are made from borosilicate glass 3.3, with core at the bottom, two 10 mm glass poles and ground clamp made of PE.