Flasks, round bottom, with three necks and standard ground joints
Lieferant: NEUBERT VOLUME GLASSWAERE
0759-0025-14
NEUB0759-0025-14EA 144 EUR
NEUB0759-0025-14
Flasks, round bottom, with three necks and standard ground joints
Kolben Rundkolben
Borosilicate glass 3.3, clear, with angled side necks.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.