Chromatography Columns, Glass Tap/Core
Lieferant: NEUBERT VOLUME GLASSWAERE
1206-1029-0200 1206-1029-2005
NEUB1206-1029-0200EA 331 EUR
NEUB1206-1029-0200 NEUB1206-1029-2005
Chromatography Columns, Glass Tap/Core
Chromatographiesäulen
  • Fried: P 0
  • NS sleeve: NS 29/32
  • Hahn: PTFE