Solid Glass Stopper
Lieferant: NEUBERT VOLUME GLASSWAERE
1641-91-3435
NEUB1641-91-3435EA 59.6 EUR
NEUB1641-91-3435
Solid Glass Stopper
Stopfen Glasstopfen
Six sided, solid glass stopper.
  • Made from Borosilicate glass 3.3
  • Six point handle