Blind cap
Lieferant: NEUBERT VOLUME GLASSWAERE
1670-10-14 1670-10-29
NEUB1670-10-14EA 16 EUR
NEUB1670-10-14 NEUB1670-10-29
Blind cap
Adapter Übergangsstücke mit Kegelschliffen
Ground cap with standard ground joints made of borosilicate glass 3.3.
  • Very good chemical resistance
  • Very good temperature resistance