Arsenic limit test apparatus
Lieferant: NEUBERT VOLUME GLASSWAERE
23900031002920
NEUB23900031002920EA 237 EUR
NEUB23900031002920
Arsenic limit test apparatus
Aufschlusssysteme Geräte zum Nachweis von Spurenelementen
This apparatus is used for the determination of the limit of arsenic.
Complete unit with four parts with springs, attachment and tension springs.
Standards: Pharmeuropa 28.2; Ph.Eur 2.4.2 method A, EP 9.4