Apparatus with flange for the determination of Arsenic
Lieferant: NEUBERT VOLUME GLASSWAERE
23900041002920
NEUB23900041002920EA 372 EUR
NEUB23900041002920
Apparatus with flange for the determination of Arsenic
Destillationsbauteile Destillations-Bausätze
Borosilicate glass 3,3.
Apparatus for the estimation of Arsenic. Complete including two springs, erlenmeyer flasks 100 ml, graduated, ST 29/32, tube with ST 29/32 and flange.
Standards: DAB 10 (Pharmeuropa 28.2 ; Ph.Eur 2.4.2 Method A )