Sublimation apparatus
Lieferant: NEUBERT VOLUME GLASSWAERE
2485-06-29
NEUB2485-06-29EA 265 EUR
NEUB2485-06-29
Sublimation apparatus
Destillationsbauteile Destillations-Bausätze
This sublimation apparatus, made of borosilicate glass 3.3, is for working under vacuum.Total height: 200 mm.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes