Eudiometer tubes
Lieferant: NEUBERT VOLUME GLASSWAERE
2591-10-509
NEUB2591-10-509EA 1340 EUR
NEUB2591-10-509
Eudiometer tubes
Flaschen Gaswaschflaschen
Eudiometer tubes with spindle tap are suitable for determining the digested behaviour of sludge and biodegradability under anaerobic conditions.
  • Made of borosilicate glass 3.3
  • With reservoir and leveling plate
  • With PVC connecting hose
Standards: According to DIN 38414 part 8.