Connection Cap, GL 25
Lieferant: NEUBERT VOLUME GLASSWAERE
6024-050-025
NEUB6024-050-025EA 5.7 EUR
NEUB6024-050-025
Connection Cap, GL 25
Verschlüsse Flaschenverschlüsse
Connection caps with hole made of celanex, without seal.
  • Drilling: 18 mm
  • Made of celanex (200°)