Plastic Olive, Curved with Screw Cap
Lieferant: NEUBERT VOLUME GLASSWAERE
6043-14-1509
NEUB6043-14-1509EA 9.7 EUR
NEUB6043-14-1509
Plastic Olive, Curved with Screw Cap
Adapter Adapter, Normschliff mit Schlaucholiven
This olive comes with perforated cap and silicone seal.
  • Cap and olive material: PP
  • Temperature resistance: 110 °C