Canada Balsam Glass Jar
Lieferant: NEUBERT VOLUME GLASSWAERE
7298-50-50
NEUB7298-50-50EA 138 EUR
NEUB7298-50-50
Canada Balsam Glass Jar
Dosen Dosen, Universal
Canada balsam glass, according to Goldschmidt, is used for microscopy and comes with a glass stirring rod.