Mobile dispensing dolly, PIG®
Lieferant: NEW PIG
DRM662
NEWPDRM662EA 1020 EUR
NEWPDRM662
Mobile dispensing dolly, PIG®
Transportwagen
This transport drum and tilts are ideal for dispensing and provides secondary containment in a closed sump.
  • Made of LDPE with poly wheels, stainless axle and nylon security strap
  • Rugged PE with UV inhibitors for indoor and outdoor use
  • Dispensing well contains minor leaks and drips from a drum tap
  • Durable and corrosion-proof
  • Ergonomically designed to minimise back strain and fatigue
  • Plastic wheels of 250 mm are excellent for use on smooth floors or concrete