PIG® HazMat chemical loose absorbent
Lieferant: NEW PIG
HA8010
NEWPHA8010EA 80 EUR
NEWPHA8010
PIG® HazMat chemical loose absorbent
Absorptionsmaterial
Loose absorbent can be sprinkled onto spills or spread into tight spaces for versatile clean-up of acids, caustics and unknown liquids.
  • Absorbs the widest range of acids, caustics and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide
  • Chemical-resistant pulp won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Pink colour is easily distinguishable to assure workers use the correct pulp during a spill emergency
  • Can be incinerated after use to reduce waste
  • Weight: 2,3 kg per bag
Ideal for difficult to reach spaces or final cleanup.
Bitte beachten To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase. If you have any questions or need samples to test, please call Technical Services.