Utility tray with grate, PIG®
Lieferant: NEW PIG
PAK123-BK
NEWPPAK123-BKEA 154.25 EUR
NEWPPAK123-BK
Utility tray with grate, PIG®
Absorptionsmaterial
Durable PE utility tray is ideal for temporary storage of leaking parts or cores awaiting degreasing, repair or pickup. It helps to save bench space as large and heavy parts like batteries, grimy cores waiting for rebuild and leaky containers can be stored safely on the floor.
  • Strong and durable, chemical resistant
  • Ribbed bottom prevents batteries and containers from resting in spilled liquids
  • Heavy-duty walls and ribbed bottom provide strength and prevent warping
  • Includes heavy-duty grate to elevate parts and containers out of deeper liquids in sump
Weight: 11,6 kg