PIG® fibreglass chemical trays
Lieferant: NEW PIG
PAK739-WH PAK740-WH PAK741-WH PAK742-WH PAK743-WH PAK744-WH
NEWPPAK739-WHEA 25.9 EUR
NEWPPAK739-WH NEWPPAK740-WH NEWPPAK741-WH NEWPPAK742-WH NEWPPAK743-WH NEWPPAK744-WH
PIG® fibreglass chemical trays
Schalen/Schüsseln/Töpfe Auffangschalen und Auffangbehälter
Catch drips and control messes with these smooth, seamless fibreglass trays.
  • Reinforced thermoset composite fibreglass resists chemicals and retains its shape, even under heavy loads
  • Smooth corners and seamless design allows trays to be steam cleaned or sterilized for repeated use
  • Molded edges safely contain minor spills or leaks from containers or oily parts
Chemical-resistant trays are easy to clean.