Spill containment pools, PIG®
Lieferant: NEW PIG
PAK925 PAK926 PAK927 PAK928
NEWPPAK925EA 125 EUR
NEWPPAK925 NEWPPAK926 NEWPPAK927 NEWPPAK928
Spill containment pools, PIG®
Absorptionsmaterial
These containment pools are ideal for quick deployment to catch the contents of blown hydraulic lines, punctured tanks, leaking containers and decontamination operations. To store our pop-up containment pool in the vehicle to be prepared for spill emergencies on the road.
  • Made of heavy-duty, 18 oz. vinyl-coated polyester provides high resistance to tears and punctures
  • Rim has foam ring that rises with level of liquid contained
  • Deploys instantly for use in an emergency; decontaminate for reuse
  • Pliable material folds easily for storage on a truck or
More Product Information