PIG® DRI loose absorbent
Lieferant: NEW PIG
PLP213-1
NEWPPLP213-1EA 22 EUR
NEWPPLP213-1
PIG® DRI loose absorbent
Absorptionsmaterial
Kick clay to the curb with this 100% recycled, silica-safe alternative. PIG DRI offers the same spreadability without the health risks.
  • Unlike clay, which is simply coated with liquid, fast-wicking cellulose-based kaolin granules begin to soak up liquid the moment they touch the spill
  • 100% recycled content is a byproduct of the paper-making process, ideal when green products are desired or required
  • Silica-free composition does not pose the health hazards associated with breathing in clay dust
  • Light-weight absorbent is easier than clay to carry and use without injury
  • Absorbs and retains oils, coolants, solvents and water
  • Can be incinerated after use to reduce waste or for fuels blending
Weight: 18,1 kg per bag
Volume bag: 30 L