PIG® acid solidifying absorbent powder
Lieferant: NEW PIG
PLP501
NEWPPLP501EA 263 EUR
NEWPPLP501
PIG® acid solidifying absorbent powder
Absorptionsmaterial
Solidifies absorbed acids and locks them into the material for easier cleanup.
  • Permanently solidifies to "lock in" absorbed liquids; won't leak or revert to liquid state when handled or placed under pressure
  • Forms a solid mass without expanding for easier, less expensive recovery and cleanup; may help you comply with EPA requirements when used to solidify waste prior to shipment or landfilling
  • High Btu value produces less than 0.1% ash content when burned; ideal for fuels blending and incineration
  • Shaker bottle makes it easy to apply absorbent directly onto the spill area
The environmentally friendly product absorbs acids and corrosives including phosphoric, sulfuric, hydrofluoric, hydrochloric and acetic acids as well as hydrazine.