PIG® GRIP-DRI absorbent granules
Lieferant: NEW PIG
PLPE260
NEWPPLPE260EA 20 EUR
NEWPPLPE260
PIG® GRIP-DRI absorbent granules
Absorptionsmaterial
Fast acting universal absorbent is chemically inert, making it compatible with almost any liquid.
  • Remains granular, even when saturated, to provide excellent retention; stands up to heavy traffic without releasing liquids
  • Fire-retardant product is ideal for use with low-flash point solvents and other flammables, and in high-temperature applications
  • Micro-granular formulation absorbs 25% more liquid than regular moler absorbents
  • Low-dust and easy to sweep away; won't leave behind an oily sheen or residue, so it helps you create cleaner, safer floors
  • Non-toxic mineral product is safe to handle and use without any risk
Instantly soaks up oils, coolants, solvents, water, acids and caustics.
Weight: 15 kg per bag
Volume bag: 30 L