Disposable drain seal
Lieferant: NEW PIG
PLRE166
NEWPPLRE166EA 177 EUR
NEWPPLRE166
Disposable drain seal
Absorptionsmaterial
Versatile product stops liquids from entering drains and causing pollution. Clay composition creates a 100 % seal, even sticking to rough, irregular surfaces like tarmac or concrete.
  • Easy to use - Simply peel of the protective plastic from one side, then press the exposed side firmly into and around the drain
  • Resistant to the most common oils and chemicals
  • Flexible at low temperatures to –20 °C and treated with a formula which prevents mould growth
Standards: Help with ADR compliance when a drain seal is required.