Lift pump for chemicals
Lieferant: NEW PIG
PMPL117
NEWPPMPL117EA 135 EUR
NEWPPMPL117
Lift pump for chemicals
Pumpen Fass/Transferpumpen
Suitable for aggressive chemicals, solvents, de-greasers and more.
  • Made of 316 stainless steel and PTFE
  • Fits standard 2" BSP or 2" NPT bung opening and works with most common drums
  • Downtube (500 mm) for use with up to 60 litre drums
  • PTFE extension hose (500 mm) for dispensing