Scoops, non sparking
Lieferant: NEW PIG
TLS290
NEWPTLS290EA 9.2 EUR
NEWPTLS290
Scoops, non sparking
Schaufeln
Non sparking scoop made from rough, resilient PP that won't rust or corrode. Ideal for handling loose absorbents, powder and contaminated soils.
  • Excellent chemical resistance for use in cleaning up loose absorbents
  • Non sparking, chemical and corrosion resistant