Maintenance wipes, heavy duty, PIG®
Lieferant: NEW PIG
WIP304 WIP306
NEWPWIP304EA 140 EUR
NEWPWIP304 NEWPWIP306
Maintenance wipes, heavy duty, PIG®
Wischtücher Wisch- und Handtücher, Einweg Wischtücher für allgemeine Anwendungen
Strong and solvent-resistant wipes made of a cellulose/polyester blend. Ideal for daily heavy duty jobs, such as wiping machines and rough parts. Also soft enough for cleaning skin or preparing surfaces.
  • Exceptional abrasion resistance and wet strength that stands up to rough surfaces, jagged edges or metal chips
  • Excellent for heavy duty maintenance jobs like machine wipedowns and degreasing parts
  • Wipes are 1/4 folded and poly wrapped for easy handling and loading
  • Low-lint construction won't shed, pill or leave behind unwanted fibres when wiping parts
  • Binder-free construction withstands solvents, degreasers and cleaning agents