Dishes with lid
Lieferant: NICKEL ELECTRO
503543
NICK503543EA 64.9 EUR
NICK503543
Dishes with lid
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Made of nickel.
  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean