Beaker tongs, stainless steel
Lieferant: NICKEL ELECTRO
7005
NICK7005EA 91.3 EUR
NICK7005
Beaker tongs, stainless steel
Zangen
These stainless steel beaker tongs feature silicone covered jaws.
  • Silicone covered sleeves reduce risk breakage when handling beakers
  • Operating temperature: −60...+180˚C
  • Hold beakers from 100 to 1000 ml