Spatula, Hayman type
Lieferant: NICKEL ELECTRO
9570
NICK9570EA 4.6 EUR
NICK9570
Spatula, Hayman type
Spatel
Stainless steel 18/8, non magnetic, bright finish.
  • Corrosion resistant and durable
  • Suitable for intermittent use up to 870 °C
This spatula has a 3 mm wide curved half moon end at the tip. This end is sharpened at the tip for cutting, separating or handling mico samples. The other end features a flat blade for handling small samples (3,5 mm wide).