Spheres for water baths, Clifton®
Lieferant: NICKEL ELECTRO
BP0368
NICKBP0368EA 223 EUR
NICKBP0368
Spheres for water baths, Clifton®
Kugeln Kunststoffkugeln
Hollow spheres are ideal for use as a blanket on the top of open tanks of water. A single layer of spheres covers 91% of the liquid surface trapping air in the gaps which adds to the thermal insulation properties. Spheres have 20 mm Ø.
  • Reduces thermal losses
  • Reduces steaming and splashing