Cleanroom wipes, BioClean Oryx™
Lieferant: Nitritex
BOWB-12 BOWB-9 BOWS-9 BOWS-9B BOWS-12
NITRBOWB-12EA 527 EUR
NITRBOWB-12 NITRBOWB-9 NITRBOWS-9 NITRBOWS-9B NITRBOWS-12
Cleanroom wipes, BioClean Oryx™
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
These cleanroom wipes are made of hydroentangled non woven polycellulose. They contain no optical brighteners or whitening agents and provide outstanding absorption properties.
  • Low particle shedding
  • Low-linting
  • High strength, smooth and durable
  • Sterile versions are gamma-irridated
Suitable for use in Class 100, ISO 5, M3.5 environments.
Colour: White.
Data accurate at time of publishing, latest test results and test methods can be found on supplier's website.
Bitte beachten Data accurate at time of publishing, latest test results and test methods can be found on supplier's website.