HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Novabiochem®

Lieferant: NOVABIOCHEM
Gefahr

8.51013.0025 8.51013.0100 8.51013.0500 8.51013.1000
01-62-0041-25EA 452 EUR
01-62-0041-25 8.51013.0100 NOVA8.51013.0500 NOVA8.51013.1000
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Novabiochem®
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)

A highly efficient coupling reagent for solid and solution phase peptide synthesis.


In comparative studies HATU has been found to give better coupling yields with less enantiomerization that HBTU, TBTU or PyBOP® [1-3]. It is particularly effective at coupling to N-alkyl amines where other coupling reagents give poor yields and is the preferred reagent for loading resins bearing secondary amino groups.

Formel: C₁₀H₁₅F₆N₆OP
Schmelzpunkt: 186 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00274639
CAS-Nummer: 148893-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR