Iod 0.02 M in THF : Pyridin : Wasser (69:22:11) (w:w:w), Novabiochem® zur DNA/RNA Synthese, oxidierendes Reagens B

Lieferant: NOVABIOCHEM
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

8.57013.4000
NOVA8.57013.4000EA 572 EUR
NOVA8.57013.4000
Iod 0.02 M in THF : Pyridin : Wasser (69:22:11) (w:w:w), Novabiochem® zur DNA/RNA Synthese, oxidierendes Reagens B
Iod
Formel: I₂
Flammpunkt: –21,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011355
CAS-Nummer: 7553-56-2
UN: 1993
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR