Anti-AIF-1/Iba1 goat polyclonal antibody
Lieferant: Novus Biologicals
NB100-1028
NOVUNB100-1028EA 780 EUR
NOVUNB100-1028
Anti-AIF-1/Iba1 goat polyclonal antibody
Antikörper
The AIF-1 / Iba1 Antibody from Novus Biologicals is a goat polyclonal antibody to AIF-1 / Iba1. This antibody reacts with human, mouse, rat. The AIF-1 / Iba1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA, Immunocytochemistry.