Anti-IREB2 Rabbit Polyclonal Antibody (DyLight® 405)
Lieferant: Novus Biologicals
NB100-1798V
NOVUNB100-1798VEA 690 EUR
NOVUNB100-1798V
Anti-IREB2 Rabbit Polyclonal Antibody (DyLight® 405)
Antikörper
Type: Primary
Antigen: IREB2 (iron-responsive element binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse